missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD21 / CR2 / C3DR Protein
A DNA sequence encoding the Mus musculus (Mouse) CD21 / CR2 / C3DR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5861-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
CD21 / CR2 / C3DR Protein | |
Research Use Only | |
Cr2 | |
Recombinant Protein | |
Yeast | |
LYGNEVSYEC DEGFYLLGEK SLQCVNDSKG HGSWSGPPPQ CLQSSPLTHC PDPEVKHGYK LNKTHSAFSH NDIVHFVCNQ GFIMNGSHLI RCHTNNTWLP GVPTCIRKAS LGCQSPSTIP NGNHTGGSIA RFPPGMSVMY SCYQGFLMAG EARLICTHEG TWSQPPPFCK EVNCSFPEDT NGIQKGFQPG KTYRFGATVT LECEDGYTLE GSPQSQCQDD SQWNPPLALC KYRRW | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |