missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD21 / CR2 / C3DR Protein
A DNA sequence encoding the Mus musculus (Mouse) CD21 / CR2 / C3DR, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Name | CD21 / CR2 / C3DR Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Cr2 |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948289
|
enQuireBio™
QP5861-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968289
|
enQuireBio™
QP5861-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958289
|
enQuireBio™
QP5861-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15978289
|
enQuireBio™
QP5861-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15988289
|
enQuireBio™
QP5861-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15998289
|
enQuireBio™
QP5861-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
CD21 / CR2 / C3DR Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
Cr2 | |
Recombinant Protein | |
E. coli | |
LYGNEVSYEC DEGFYLLGEK SLQCVNDSKG HGSWSGPPPQ CLQSSPLTHC PDPEVKHGYK LNKTHSAFSH NDIVHFVCNQ GFIMNGSHLI RCHTNNTWLP GVPTCIRKAS LGCQSPSTIP NGNHTGGSIA RFPPGMSVMY SCYQGFLMAG EARLICTHEG TWSQPPPFCK EVNCSFPEDT NGIQKGFQPG KTYRFGATVT LECEDGYTLE GSPQSQCQDD SQWNPPLALC KYRRW | |
Greater than 90% as determined by SDS-PAGE. |