missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD40 Protein

Product Code. 15906908
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15906908

Brand: enQuireBio™ QP113342ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the CD40 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 21939
Name CD40 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol Cd40
Product Type Recombinant Protein
Cross Reactivity Mouse
Species Baculovirus
Protein Tag His
Sequence LGQCVTCSDK QYLHDGQCCD LCQPGSRLTS HCTALEKTQC HPCDSGEFSA QWNREIRCHQ HRHCEPNQGL RVKKEGTAES DTVCTCKEGQ HCTSKDCEAC AQHTPCIPGF GVMEMATETT DTVCHPCPVG FFSNQSSLFE KCYPWTSCED KNLEVLQKGT SQTNVICGLK SRMRHHHHHH
Buffer CD40 protein solution (1.0 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.