missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD68 / Macrosialin Protein

A DNA sequence encoding the Mus musculus (Mouse) CD68 / Macrosialin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5793-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15917809

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

NP_001277987.1
CD68 / Macrosialin Protein
Research Use Only
Cd68
Recombinant Protein
Yeast
DCPHKKAVTL LPSFTMTPTA TESTASPTTS HRPTTTSHGN VTVHTSSGPT TVTHNPATTT SHGNATISHA TVSPTTNGTA TSPRSSTVGP HPGPPPPSPS PRSKGALGNY TWANGSQPCV QLQAQIQIRI LYPIQGGRKA WGISVLNPNK TKVQGGCDGT HPHLSLSFPY GQLTFGFKQD LHQSPSTVYL DYMAVEYNVS FPQAAQWTFM AQNSSLRELQ APLGQSFCCG NASIVLSPAV HLDLLSLRLQ AAQLPDKGHF GPCFSCNRDQ S
Greater than 90% as determined by SDS-PAGE.
12514
200 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions