missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD68 / Macrosialin Protein
A DNA sequence encoding the Mus musculus (Mouse) CD68 / Macrosialin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5793-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001277987.1 | |
CD68 / Macrosialin Protein | |
Research Use Only | |
Cd68 | |
Recombinant Protein | |
Yeast | |
DCPHKKAVTL LPSFTMTPTA TESTASPTTS HRPTTTSHGN VTVHTSSGPT TVTHNPATTT SHGNATISHA TVSPTTNGTA TSPRSSTVGP HPGPPPPSPS PRSKGALGNY TWANGSQPCV QLQAQIQIRI LYPIQGGRKA WGISVLNPNK TKVQGGCDGT HPHLSLSFPY GQLTFGFKQD LHQSPSTVYL DYMAVEYNVS FPQAAQWTFM AQNSSLRELQ APLGQSFCCG NASIVLSPAV HLDLLSLRLQ AAQLPDKGHF GPCFSCNRDQ S | |
Greater than 90% as determined by SDS-PAGE. |
12514 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |