missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CES1G Protein

A cDNA sequence encoding the CES1G was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Gene ID (Entrez) 12623
Name CES1G Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol Ces1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15927128
View Documents Promotion Details
enQuireBio™
QP11393-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15907128
View Documents Promotion Details
enQuireBio™
QP11393-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15917128
View Documents Promotion Details
enQuireBio™
QP11393-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

12623
Research Use Only
Ces1
Mouse
His
CES1G protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
CES1G Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
ADPHPSLPPV VHTVHGKVLG KYVTLEGFSQ PVAVFLGVPF AKPPLGSLRF APPEPAEPWS FVKHTTSYPP LCYQNPEAAL RLAELFTNQR KIIPHKFSED CLYLNIYTPA DLTQNSRLPV MVWIHGGGLV IDGASTYDGV PLAVHENVVV VVIQYRLGIW GFFSTEDEHS RGNWGHLDQV AALHWVQDNI ANFGGNPGSV TIFGESAGGE SVSVLVLSPL AKNLFHRAIA QSSVIFNPCL FGRAARPLAK KIAALAGCKT TTSAAMVHCL RQKTEDELLE VSLKMKFGTV DFLGDPRESY PFLPTVIDGV LLPKAPEEIL AEKSFNTVPY MVGINKHEFG WIIPMFLDFP LSERKLDQKT AASILWQAYP ILNISEKLIP AAIEKYLGGT EDPATMTDLF LDLIGDIMFG VPSVIVSRSH RDAGAPTYMY EYQYRPSFVS DDRPQELLGD HADELFSVWG APFLKEGASE EEINLSKMVM KFWANFARNG NPNGEGLPHW PEYDQKEGYL QIGVPAQAAH RLKDKEVDFW TELRAKETAE RSSHREHVEL HHHHHH
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications