missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Chymotrypsin-like elastase family member 2A Protein

A DNA sequence encoding the Mus musculus (Mouse) Chymotrypsin-like elastase family member 2A, was expressed in the hosts and tags indicated.

245.00€ - 2645.00€

Specifications

Accession Number NP_031945.1
Gene ID (Entrez) 13706
Name Chymotrypsin-like elastase family member 2A Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15927979
View Documents Promotion Details
enQuireBio™
QP5817-YE-100UG
100 μg
770.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15967979
View Documents Promotion Details
enQuireBio™
QP5817-YE-500UG
500 μg
2195.00€
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15937979
View Documents Promotion Details
enQuireBio™
QP5817-YE-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15957979
View Documents Promotion Details
enQuireBio™
QP5817-YE-200UG
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15977979
View Documents Promotion Details
enQuireBio™
QP5817-YE-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15947979
View Documents Promotion Details
enQuireBio™
QP5817-YE-1MG
1 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_031945.1
Chymotrypsin-like elastase family member 2A Protein
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
13706
Research Use Only
Cela2a
Recombinant Protein
Yeast
VVGGQEATPN TWPWQVSLQV LSSGRWRHNC GGSLVANNWV LTAAHCLSNY QTYRVLLGAH SLSNPGAGSA AVQVSKLVVH QRWNSQNVGN GYDIALIKLA SPVTLSKNIQ TACLPPAGTI LPRNYVCYVT GWGLLQTNGN SPDTLRQGRL LVVDYATCSS ASWWGSSVKS SMVCAGGDGV TSSCNGDSGG PLNCRASNGQ WQVHGIVSFG SSLGCNYPRK PSVFTRVSNY IDWINSVMAR N
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications