missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cyclophilin B Protein
A cDNA sequence encoding the Cyclophilin B was constructed and used to recombinantly synthesize the protein.
137.00€ - 7875.00€
Specifications
Gene ID (Entrez) | 19035 |
---|---|
Name | Cyclophilin B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Ppib |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934278
|
enQuireBio™
QP10569-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914278
|
enQuireBio™
QP10569-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924278
|
enQuireBio™
QP10569-1MG |
1 mg |
7875.00€
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
19035 | |
Research Use Only | |
Ppib | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNDKKKG PKVTVKVYFD LQIGDESVGR VVFGLFGKTV PKTVDNFVAL ATGEKGFGYK NSKFHRVIKD FMIQGGDFTR GDGTGGKSIY GERFPDENFK LKHYGPGWVS MANAGKDTNG SQFFITTVKT SWLDGKHVVF GKVLEGMDVV RKVESTKTDSRDKPLKDVII VDSGKIEVEK PFAIAKE | |
Greater than 95.0% as determined by SDS-PAGE. |
Cyclophilin B Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 1,000 nmol/min/mg, and is defined as the amount of enzyme that cleaves 1nmole of suc-AAFP-PNA per minute at 37C in Tris€“HCl pH 8.0 using chymotrypsin. | |
Mouse | |
His | |
Cyclophilin B protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |