missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Ephrin B3 / EFNB3 Protein

A DNA sequence encoding the Mus musculus (Mouse) Ephrin B3 / EFNB3, was expressed in the hosts and tags indicated.

245.00€ - 3405.00€

Specifications

Accession Number NP_031937.1
Gene ID (Entrez) 13643
Name Ephrin B3 / EFNB3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15928999
View Documents Promotion Details
enQuireBio™
QP5967-EC-100UG
100 μg
770.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15948999
View Documents Promotion Details
enQuireBio™
QP5967-EC-1MG
1 mg
3405.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15938999
View Documents Promotion Details
enQuireBio™
QP5967-EC-10UG
10 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15958999
View Documents Promotion Details
enQuireBio™
QP5967-EC-200UG
200 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15968999
View Documents Promotion Details
enQuireBio™
QP5967-EC-500UG
500 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15978999
View Documents Promotion Details
enQuireBio™
QP5967-EC-50UG
50 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_031937.1
Ephrin B3 / EFNB3 Protein
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
13643
Research Use Only
Efnb3
Recombinant Protein
E. coli
LSLEPVYWNS ANKRFQAEGG YVLYPQIGDR LDLLCPRARP PGPHSSPSYE FYKLYLVEGA QGRRCEAPPA PNLLLTCDRP DLDLRFTIKF QEYSPNLWGH EFRSHHDYYI IATSDGTREG LESLQGGVCL TRGMKVLLRV GQSPRGGAVP RKPVSEMPME RDRGAAHSAE PGRDTIPGDP SSNATSRGAE GPLPPPSMPA
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications