missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse GITR Ligand/TNFSF18 (aa 47-173)
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS.
Unit Size:
25µg
Description
Measured by its ability to stimulate mouse T cell proliferation in the presence of anti-CD3.
The ED50 for this effect is 0.02-0.3 μg/mL in the presence of 10 μg/mL of a cross-linking antibody, Mouse Anti-polyHistidine Monoclonal Antibody (Catalog # MAB050 ).
Specifications
Specifications
| Accession Number | AAO89011 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 8995 |
| Molecular Weight (g/mol) | M.W. (Observed): 16-26 kDa, reducing conditions; M.W. (theoretical): 16 kDa |
| Name | GITR Ligand/TNFSF18 aa 47-173 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse GITR Ligand/TNFSF18 protein Thr47-Ser173, with an N-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction