missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GSTP2 Protein
A cDNA sequence encoding the GSTP2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12063-HIS-10ug
Additional Details : Weight : 0.01000kg
Specifications
14869 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Gstp2 | |
Mouse | |
His | |
GSTP2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
GSTP2 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPPYTIV YFPSPGRCEA MRMLLADQGQ SWKEEVVTID TWMQGLLKPT CLYGQLPKFE DGDLTLYQSN AILRHLGRSL GLYGKNQREA AQVDMVNDGV EDLRGKYGTM IYRNYENGKN DYVKALPGHL KPFETLLSQN QGGKAFIVGD QISFADYNLL DLLLIHQVLA PGCLDNFPLL SAYVARLSAR PKIKAFLSSP EHVNRPINGN GKQ |