missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL 18 Protein

A cDNA sequence encoding the IL 18 was constructed and used to recombinantly synthesize the protein.

137.00€ - 1815.00€

Specifications

Gene ID (Entrez) 16173
Name IL 18 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol Il18
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15930279
View Documents Promotion Details
enQuireBio™
QP12394-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15920279
View Documents Promotion Details
enQuireBio™
QP12394-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15910279
View Documents Promotion Details
enQuireBio™
QP12394-0.1MG
100 μg
1815.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

16173
Research Use Only
Il18
Mouse
Untagged
The IL 18 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4).
IL 18 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MNFGRLHCTT AVIRNINDQV LFVDKRQPVF EDMTDIDQSA SEPQTRLIIY MYKDSEVRGL AVTLSVKDSK MSTLSCKNKI ISFEEMDPPE NIDDIQSDLI FFQKRVPGHN KMEFESSLYE GHFLACQKED DAFKLILKKK DENGDKSVMF TLTNLHQS
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications