missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-6 Protein
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.02-0.06 ng/mL.
Specifications
Specifications
| Accession Number | P08505 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate and EDTA. |
| Gene ID (Entrez) | 3569 |
| Molecular Weight (g/mol) | M.W. (theoretical): 21.8 kDa |
| Name | IL-6 |
| Quantity | 25 μg |
| Source | E. coli-derived mouse IL-6 protein Phe25-Thr211, with an N-terminal Met |
| Storage Requirements | Do not freeze. 6 months from date of receipt, 2°C to 8°C as supplied. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction