missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse IL10 / Interleukin-10 Protein

Product Code. 15926239
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
This item is not returnable. View return policy

Product Code. 15926239

Brand: enQuireBio™ QP5490500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_034678.1
Gene ID (Entrez) 16153
Name IL10 / Interleukin-10 Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol IL10
Biological Activity Shows bioactivity at a concentration of 5 ng/mL in the MC/9 cell proliferation assay. The bioactivity acceptance criteria is set at 2.0 x 10^5 units/mg.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MSRGQYSRED NNCTHFPVGQ SHMLLELRTA FSQVKTFFQT KDQLDNILLT DSLMQDFKGY LGCQALSEMI QFYLVEVMPQ AEKHGPEIKE HLNSLGEKLK TLRMRLRRCH RFLPCENKSK AVEQVKSDFN KLQDQGVYKA MNEFDIFINC IEAYMMIKMK S
Buffer 10 mM sodium phosphate, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.