missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LDHA Protein
A cDNA sequence encoding the LDHA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10743-2ug
Additional Details : Weight : 0.01000kg
Specifications
16828 | |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Ldha | |
Recombinant Protein | |
Insect | |
MATLKDQLIV NLLKEEQAPQ NKITVVGVGA VGMACAISIL MKDLADELAL VDVMEDKLKG EMMDLQHGSL FLKTPKIVSS KDYCVTANSK LVIITAGARQ QEGESRLNLV QRNVNIFKFI IPNIVKYSPH CKLLIVSNPV DILTYVAWKI SGFPKNRVIG SGCNLDSARF RYLMGERLGV HALSCHGWVL GEHGDSSVPV WSGVNVAGVS LKSLNPELGT DADKEQWKEV HKQVVDSAYE VIKLKGYTSW AIGLSVADLA ESIMKNLRRV HPISTMIKGL YGINEDVFLS VPCILGQNGI SDVVKVTLTP EEEARLKKSA DTLWGIQKEL QFLEHHHHHH |
LDHA Protein | |
2 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 250 units/mg, and is defined as the Amount of enzyme that convert 1.0 umole of pyruvate to L-lactate and per minute at pH 7.5 at 37C. | |
Mouse | |
His | |
LDHA protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |