missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MCP 1 Protein

Product Code. 15975148
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15975148

Brand: enQuireBio™ QP1078110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the MCP 1 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 20296
Name MCP 1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol Ccl2
Biological Activity The biological activity was determined by calculating its ability to chemoattract Balb/C mouse spleen MNCs at 1.0-20.0 ng/ml.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MQVPVMLLGL LFTVAGWSIH VLAQPDAVNA PLTCCYSFTS KMIPMSRLES YKRITSSRCP KEAVVFVTKL KREVCADPKK EWVQTYIKNL DRNQMRSEPT TLFKTASALR SSAPLNVKLT RKSEANASTT FSTTTSSTSV GVTSVTVN
Buffer The protein was lyophilized with no additives.
Purity or Quality Grade Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.