missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Midkine Protein

Product Code. 15925168
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15925168

Brand: enQuireBio™ QP107861mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Midkine was constructed and used to recombinantly synthesize the protein.

Specifications

Name Midkine Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Fully biologically active when compared to standard. Determined by its ability to chemoattract human neutrophils using a concentration range of 10-100 ng/ml corresponding to a specific activity of 10,000-100,000IU/mg.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence VAKKKEKVKK GSECSEWTWG PCTPSSKDCG MGFREGTCGA QTQRVHCKVP CNWKKEFGAD CKYKFESWGA CDGSTGTKAR QGTLKKARYN AQCQETIRVT KPCTSKTKSK TKAKKGKGKD
Buffer Lyophilized from a 0.2um filtered concentrated solution in PBS, pH 7.4.
Purity or Quality Grade Greater than 95.0% as determined by HPLC and SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.