missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse PARK7 Protein

A cDNA sequence encoding the PARK7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12963-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932089

  • 3740.00€ / 1mg

Estimated Shipment: 01-08-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

PARK7 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
PARK7 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMASKRAL VILAKGAEEM ETVIPVDVMR RAGIKVTVAG LAGKDPVQCS RDVMICPDTS LEDAKTQGPY DVVVLPGGNL GAQNLSESPM VKEILKEQES RKGLIAAICA GPTALLAHEV GFGCKVTTHP LAKDKMMNGS HYSYSESRVE KDGLILTSRG PGTSFEFALA IVEALVGKDM ANQVKAPLVL KD
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions