missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Resistin / ADSF / RETN Protein

Recombinant Protein

Brand:  enQuireBio™ QP5437-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15916129

  • 263.00€ / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001191888.1
Resistin / ADSF / RETN Protein
Research Use Only
RETN
Recombinant Protein
E. coli
MSSMPLCPID EAIDKKIKQD FNSLFPNAIK NIGLNCWTVS SRGKLASCPE GTAVLSCSCG SACGSWDIRE EKVCHCQCAR IDWTAARCCK LQVAS
Greater than 95% as determined by SDS-PAGE.
57264
25 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Mouse Resistin / ADSF / RETN induces IL-8 production in human PBMCs detected via ELISA (typical ED50 is 400-600 ng/ml).
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions