missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Resistin / ADSF / RETN Protein
Recombinant Protein
Brand: enQuireBio™ QP5437-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001191888.1 | |
Resistin / ADSF / RETN Protein | |
Research Use Only | |
RETN | |
Recombinant Protein | |
E. coli | |
MSSMPLCPID EAIDKKIKQD FNSLFPNAIK NIGLNCWTVS SRGKLASCPE GTAVLSCSCG SACGSWDIRE EKVCHCQCAR IDWTAARCCK LQVAS | |
Greater than 95% as determined by SDS-PAGE. |
57264 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse Resistin / ADSF / RETN induces IL-8 production in human PBMCs detected via ELISA (typical ED50 is 400-600 ng/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |