missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse S100A6 Protein

A cDNA sequence encoding the S100A6 was constructed and used to recombinantly synthesize the protein.

276.00€ - 745.00€

Specifications

Gene ID (Entrez) 20200
Name S100A6 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol S100a6
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15913439
View Documents Promotion Details
enQuireBio™
QP13397-2UG
2 μg
276.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15923439
View Documents Promotion Details
enQuireBio™
QP13397-5UG
5 μg
414.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15903439
View Documents Promotion Details
enQuireBio™
QP13397-10UG
10 μg
745.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

20200
Research Use Only
S100a6
Mouse
Untagged
The protein was lyophilized from a concentrated solution (1 mg/ml) containing no additives.
S100A6 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MACPLDQAIG LLVAIFHKYS GKEGDKHTLS KKELKELIQK ELTIGSKLQD AEIARLMDDL DRNKDQEVNF QEYVAFLGAL ALIYNEALK
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications