missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse sRANKL Protein

A cDNA sequence encoding the sRANKL was constructed and used to recombinantly synthesize the protein.

137.00€ - 7090.00€

Specifications

Name sRANKL Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The ED50, as measured by its ability to induce osteoclast differentiation of RAW 264.7 mouse monocyte/macrophage cells, is less than 2ng/ml.
Product Type Recombinant Protein
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15935418
View Documents Promotion Details
enQuireBio™
QP10869-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915418
View Documents Promotion Details
enQuireBio™
QP10869-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925418
View Documents Promotion Details
enQuireBio™
QP10869-1MG
1 mg
7090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

sRANKL Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
Lys158-Asp316, with an N-terminal Met, Accession # Q3TWY5
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
The ED50, as measured by its ability to induce osteoclast differentiation of RAW 264.7 mouse monocyte/macrophage cells, is less than 2ng/ml.
Mouse
Untagged
sRANKL protein was lyophilized from a 0.2microm filtered solution in NaH2PO4, NaCl and EDTA.
Videos
SDS
Documents

Documents

Product Certifications