missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse Thrombopoietin/THPO Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1mg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
Thrombopoietin Protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human MO7e cells is less than 1.0 ng/mL, corresponding to a specific activity of >1.0 x 10^6 IU/mg.
Specifications
Specifications
| Accession Number | P40226, P40226, P40226 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, with 5% Trehalose. |
| Gene ID (Entrez) | 7066 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 18.7 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of Thrombopoietin/THPO as determined by LAL method. |
| Gene Alias | Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDFC-mpl ligand, ML, MPL ligand, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, thrombopoietin, thrombopoietin nirs variant 1, TPOMKCSF |
| Gene Symbol | THPO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction