missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse TIM-3 hIgG-His Protein

Product Code. 18607818 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
50 μg
500 μg
Unit Size:
100µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 18607818

Brand: Novus Biologicals™ NBP307123500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4) and 10% Glycerol
Gene ID (Entrez) 84868
Molecular Weight (g/mol) MolecularWeight-theroretical: 46.3 kDa
Quantity 500 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less Than1 EU / 1 μg of protein (determined by LAL method)
Gene Alias CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Gene Symbol HAVCR2
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Form Purified
Research Category Cancer, Immunology, Innate Immunity, Neuroscience
Purity or Quality Grade >95%, by SDS-PAGE
Protein TIM-3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.