missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TNFR Protein
A cDNA sequence encoding the TNFR was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Gene ID (Entrez) | 21937 |
---|---|
Name | TNFR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Tnfrsf1a |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945468
|
enQuireBio™
QP10885-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935468
|
enQuireBio™
QP10885-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925468
|
enQuireBio™
QP10885-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
21937 | |
Research Use Only | |
Tnfrsf1a | |
Recombinant Protein | |
E. coli | |
IHPSGVTGLV PSLGDREKRD SLCPQGKYVH SKNNSICCTK CHKGTYLVSD CPSPGRDTVC RECEKGTFTA SQNYLRQCLS CKTCRKEMSQ VEISPCQADK DTVCGCKENQ FQRYLSETHF QCVDCSPCFN GTVTIPCKET QNTVCNCHAG FFLRESECVP CSHCKKNEEC MKLCLPPPLA NVTNPQDSGT A | |
Greater than 97.0% as determined by SDS-PAGE. |
TNFR Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by its ability to inhibit the TNF-? mediated cytotoxicity in the L-929 cells is less than 1.0 microg/ml, corresponding to a specific activity of > 1000 IU/mg in the presence of 0.1 ng/mL of rMuTNF-a. | |
Mouse | |
Untagged | |
TNFR protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |