missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse TREM2 Protein
A DNA sequence encoding the Mus musculus (Mouse) TREM2, was expressed in the hosts and tags indicated.
364.00€ - 3745.00€
Specifications
Accession Number | NP_001259007.1 |
---|---|
Gene ID (Entrez) | 83433 |
Name | TREM2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995589
|
enQuireBio™
QP4435-10UG |
10 μg |
364.00€
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925599
|
enQuireBio™
QP4435-20UG |
20 μg |
680.00€
20µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975599
|
enQuireBio™
QP4435-MA-100UG |
100 μg |
1690.00€
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915599
|
enQuireBio™
QP4435-200UG |
200 μg |
1390.00€
200µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935599
|
enQuireBio™
QP4435-500UG |
500 μg |
2360.00€
500µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905599
|
enQuireBio™
QP4435-1MG |
1 mg |
3745.00€
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985599
|
enQuireBio™
QP4435-MA-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
NP_001259007.1 | |
TREM2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
83433 | |
Research Use Only | |
Trem2 | |
Recombinant Protein | |
Mammal | |
LNTTVLQGMA GQSLRVSCTY DALKHWGRRK AWCRQLGEEG PCQRVVSTHG VWLLAFLKKR NGSTVIADDT LAGTVTITLK NLQAGDAGLY QCQSLRGREA EVLQKVLVEV LEDPLDDQDA GDLWVPEESS SFEGAQVEHS TSRNQETSFP PTS | |
Greater than 90% as determined by SDS-PAGE. |