missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Multi Species PRKAR1A Protein

Product Code. 15922639
Click to view available options
Quantity:
1 μg
100 μg
3 μg
Unit Size:
100µg
1µg
3µg
This item is not returnable. View return policy

Product Code. 15922639

Brand: enQuireBio™ QP131431ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PRKAR1A was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 5573
Name Multi Species PRKAR1A Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol PRKAR1A
Product Type Recombinant Protein
Cross Reactivity Multi Species
Species E. coli
Protein Tag Untagged
Sequence MESGSTAASE EARSLRECEL YVQKHNIQAL LKDSIVQLCT ARPERPMAFL REYFERLEKE EAKQIQNLQK AGTRTDSRED EISPPPPNPV VKGRRRRGAI SAEVYTEEDA ASYVRKVIPK DYKTMAALAK AIEKNVLFSH LDDNERSDIF DAMFSVSFIA GETVIQQGDE GDNFYVIDQG ETDVYVNNEW ATSVGEGGSF GELALIYGTP RAATVKAKTN VKLWGIDRDS YRRILMGSTL RKRKMYEEFL SKVSILESLD KWERLTVADA LEPVQFEDGQ KIVVQGEPGD EFFIILEGSA AVLQRRSENE EFVEVGRLGP SDYFGEIALL MNRPRAATVV ARGPLKCVKL DRPRFERVLG PCSDILKRNI QQYNSFVSLS V
Buffer PKA regulatory subunit I a is supplied in 50% glycerol.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.