missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Oropouche virus nucleoProtein
Recombinant Protein
Brand: enQuireBio™ QP10488-100ug
Additional Details : Weight : 0.01000kg
Specifications
Oropouche virus nucleoProtein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
15mM Tris-Hcl, 225mM NaCl, 20% glycerol, pH7.5. |
100 μg | |
Untested | |
Oropouche virus, strain TRVL9760 | |
Native | |
>90% as determined by SDS-PAGE. |