missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other 1F8 Chagas Protein

Product Code. 15995568
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15995568

Brand: enQuireBio™ QP109195ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the 1F8 Chagas was constructed and used to recombinantly synthesize the protein.

Specifications

Name 1F8 Chagas Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag His
Sequence MGACGSKGST SDKGLASDKD GKKAKDRKEA WERIRQAIPR EKTAEAKQRR IELFKKFDKN ETGKLCYDEV HSGCLEVLKL DEFTPRVRDI TKRAFDKARA LGSKLENKGS EDFVEFLEFR LMLCYIYDFF ELTVMFDEID ASGNMLVDEE ELKRAVPKLE AWGAKVEDPA ALFKELDKNG TGSVTFDEFA AWASAVKLDA DGDPDNVPES A
Buffer 1F8 Chagas antigen was lyophilized from 20mM Hepes, 300mM NaCl, pH 8.0, and varying 284mM imidazole.
Purity or Quality Grade Protein is >90% pure as determined by SDS PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.