missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Antide Protein
A cDNA sequence encoding the Antide was constructed and used to recombinantly synthesize the protein.
137.00€ - 745.00€
Specifications
Name | Antide Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15915918
|
enQuireBio™
QP11028-1mg |
1 mg |
137.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15935918
|
enQuireBio™
QP11028-5mg |
5 mg |
212.00€
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15925918
|
enQuireBio™
QP11028-25mg |
25 mg |
745.00€
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Antide Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The Antide peptide was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Ac-D-2-Nal-D-Phe(4-CL)-3-D-Pal-Ser-Lys(nicotinoyl)-D-Lys(nicotinoyl)-Leu-Lys(Isopropyl)-Pro-D-Ala-Nh2 | |
Greater than 98.0% as determined by RP-HPLC. |