missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Batroxobin Protein

Product Code. p-7148949
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15906248

Brand: enQuireBio™ QP1113010ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Batroxobin was constructed and used to recombinantly synthesize the protein.

Specifications

Name Batroxobin Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Sequence MVLIRVIANL LILQVSYAQK SSELVIGGDE CDINEHPFLA FMYYSPRYFC GMTLINQEWV LTAAHCNRRF MRIHLGKHAG SVANYDEVVR YPKEKFICPN KKKNVITDKD IMLIRLDRPV KNSEHIAPLS LPSNPPSVGS VCRIMGWGAI TTSEDTYPDV PHCANINLFN NTVCREAYNG LPAKTLCAGV LQGGIDTCGG DSGGPLICNG QFQGILSWGS DPCAEPRKPA FYTKVFDYLP WIQSIIAGNK TATCP
Buffer The Batroxobin protein was lyophilized from a concentrated (1 mg/ml) solution with no additives.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.