missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Eledoisin Protein
Click to view available options
Quantity:
5 mg
20 mg
100 mg
Unit Size:
100mg
20mg
5mg
Specifications
Specifications
| Name | Eledoisin Protein |
| Quantity | 5 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | The protein (1 mg/ml) was lyophilized with no additives. |
| Purity or Quality Grade | Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction