missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other GHRP 6 Protein

A cDNA sequence encoding the GHRP 6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11969-25mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978958

Quantity Price
1 171.64€ / 25mg
10 161.27€ / 25mg
Estimated Shipment
01-04-2021
Add to basket


Description

Description

Specifications

Specifications

GHRP 6 Protein
25 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The GHRP-6 peptide was lyophilized with no additives.
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
His-D-Trp-Ala-Trp-D-Phe-Lys-NH2
Videos
SDS
Documents