missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GLC8 Yeast Protein
A cDNA sequence encoding the GLC8 Yeast was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11981-50ug
Additional Details : Weight : 0.01000kg
Specifications
GLC8 Yeast Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMGGI LKNPLALSPE QLAQQDPETL EEFRRQVYEN TQKNAKLTSH KRNIPGLDNT KEEGEIIGTS STFLPKDTLS LKHEQDMLAK MTPEERVQWN QRNLAENEIT KKQFQDIHID EPKTPYQGAV DPHGEYYRVD DDEDEDNSDK KPCQVANDDI DDLSLGEPEF EIKENKQPDF ETNDENDEDS PEARHKKFEE MRKKHYDVRA IFNKKSREAL KDEDEDEDDS TTKEP | |
Greater than 95% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
The protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT, and 10% glycerol. |