missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Gliadin Alpha Wheat Protein

A cDNA sequence encoding the Gliadin Alpha Wheat was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11983-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988998

  • 1815.00€ / 1mg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Gliadin Alpha Wheat Protein
Research Use Only
Recombinant Protein
E. coli
MVRVPVPQLQPQNPSQQQPQEQVPLVQQQQFPGQQQPFPPQQPYPQPQPFPSQQPYLQLQPFPQPQLPYPQPQLPYPQPQLPYPQPQPFRPQQPYPQSQPQYSQPQQPISQQQQQQQQQQQQKQQQQQQQQILQQILQQQLIPCRDVVLQQHSIAYGSSQVLQQSTYQLVQQLCCQQLWQIPEQSRCQAIHNVVHAIILHQQQQQQQQQQQQPLSQVSFQQPQQQYPSGQGSFQPSQQNPQAQGSVQPQQLPQFEEIRNLALETLPAMCNVYIPPYCTIAPVGIFGTNYRHHHHHH
Protein is >90% pure.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
Gliadin Alpha protein solution (1 mg/ml) in 10mM Tris-HCl pH 7.2.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions