missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other GroES Protein
A cDNA sequence encoding the GroES was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12052-1mg
Additional Details : Weight : 0.01000kg
Specifications
GroES Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The GroES protein (1 mg/ml) contains 25mM Tris-HCl buffer (pH 7.5), 100mM NaCl, 5mM DTT and 10% Glycerol. |
Greater than 95.0% as determined by analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MNIRPLHDRV IVKRKEVETK SAGGIVLTGS AAAKSTRGEV LAVGNGRILE GEVKPLDVKVGDIVIFNDG YGVKSEKIDN EEVLIMSESD ILAIVEA |