missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HEV Mosaic Protein
A cDNA sequence encoding the HEV Mosaic was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12221-500ug
Additional Details : Weight : 0.01000kg
Specifications
HEV Mosaic Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, 1mM EDTA, 0.5M urea and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
HEV Mosaic protein is >95% pure as determined by 10% PAGE (coomassie staining). |