missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other HRP Protein
Click to view available options
Quantity:
20 mg
60 mg
1 g
Unit Size:
1g
20mg
60mg
Specifications
Specifications
| Name | HRP Protein |
| Quantity | 20 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | Greater than 268 U/mg (25C, guaiacol as the hydrogen donor, pH 7 and H2O2 as substrates). |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | Non-species Specific |
| Protein Tag | Untagged |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction