missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other HSA Protein

A cDNA sequence encoding the HSA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12314-50mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15920029

  • 212.00€ / 50mg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

213
50 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
P. pastoris
RGVFRRDAHK SEVAHRFKDL GEENFKALVL IAFAQYLQQC PFEDHVKLVN EVTEFAKTCV ADESAENCDK SLHTLFGDKL CTVATLRETY GEMADCCAKQ EPERNECFLQ HKDDNPNLPR LVRPEVDVMC TAFHDNEETF LKKYLYEIAR RHPYFYAPEL LFFAKRYKAA FTECCQAADK AACLLPKLDE LRDEGKASSA KQRLKCASLQ KFGERAFKAW AVARLSQRFP KAEFAEVSKL VTDLTKVHTE CCHGDLLECA DDRADLAKYI CENQDSISSK LKECCEKPLL EKSHCIAEVE NDEMPADLPS LAADFVESKD VCKNYAEAKD VFLGMFLYEY ARRHPDYSVV LLLRLAKTYE TTLEKCCAAA DPHECYAKVF DEFKPLVEEP QNLIKQNCEL FEQLGEYKFQ NALLVRYTKK VPQVSTPTLV EVSRNLGKVG SKCCKHPEAK RMPCAEDYLS VVLNQLCVLH EKTPVSDRVT KCCTESLVNR RPCFSALEVD ETYVPKEFNA ETFTFHADIC TLSEKERQIK KQTALVELVK HKPKATKEQL KAVMDDFAAF VEKCCKADDK ETCFAEEGKK LVAASQAALG LHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
HSA Protein
Research Use Only
ALB
Other
Untagged
HSA protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions