missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other Lysostaphin Protein

A cDNA sequence encoding the Lysostaphin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10778-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935138

  • 125.00€ / 1mg

Estimated Shipment: 06-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Lysostaphin Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MKKTKNNYYT RPLAIGLSTF ALASIVYGGI QNETHASEKS NMDVSKKVAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKAPV ENTAEVETSK APVENTAEVE TSKAPVENTA EVETSKAPVE NTAEVETSKA PVENTAEVET SKAPVENTAE VETSKAPVEN TAEVETSKAP VENTAEVETS KAPVENTAEV ETSKAPVENT AEVETSKALV QNRTALRAAT HEHSAQWLNN YKKGYGYGPY PLGINGGMHY GVDFFMNIGT PVKAISSGKI VEAGWSNYGG GNQIGLIEND GVHRQWYMHL SKYNVKVGDY VKAGQIIGWS GSTGYSTAPH LHFQRMVNSF SNSTAQDPMP FLKSAGYGKA GGTVTPTPNT GWKTNKYGTL YKSESASFTP NTDIITRTTG PFRSMPQSGV LKAGQTIHYD EVMKQDGHVW VGYTGNSGQR IYLPVRTWNK STNTLGVLWG TIK
97% as determined by RP-HPLC.
Research Use Only
lss
Other
Untagged
The protein was lyophilized without any additives.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions