Learn More
enQuireBio™ Recombinant other MT I Protein
Recombinant Protein
Brand: enQuireBio™ QP12745-100mg
Additional Details : Weight : 0.01000kg
Specifications
MT I Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein (1 mg/ml) was lyophilized with no additives. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by RP-HPLC. |