missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other MutY Protein

A cDNA sequence encoding the MutY was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12757-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931449

  • 1525.00€ / 50µg

Estimated Shipment: 26-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

MutY Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
His
The MutY solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl and 20% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMQASQFS AQVLDWYDKY GRKTLPWQID KTPYKVWLSE VMLQQTQVAT VIPYFERFMA RFPTVTDLAN APLDEVLHLW TGLGYYARAR NLHKAAQQVA TLHGGKFPET FEEVAALPGV GRSTAGAILS LSLGKHFPIL DGNVKRVLAR CYAVSGWPGK KEVENKLWSL SEQVTPAVGV ERFNQAMMDL GAMICTRSKP KCSLCPLQNG CIAAANNSWA LYPGKKPKQT LPERTGYFLL LQHEDEVLLA QRPPSGLWGG LYCFPQFADE ESLRQWLAQR QIAADNLTQL TAFRHTFSHF HLDIVPMWLP VSSFTGCMDE GNALWYNLAQ PPSVGLAAPV ERLLQQLRTG APV
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions