missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other PROK Tritirachium album Protein

A cDNA sequence encoding the PROK Tritirachium album was constructed and used to recombinantly synthesize the protein.

137.00€ - 1450.00€

Specifications

Name PROK Tritirachium album Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol PROK
Biological Activity 34 Units/mg protein. One unit is defined as the amount of enzyme that will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 mol tyrosine per min at 37C, pH 7.5 (color by Folin-Ciocalteu reagent).
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15935358
View Documents Promotion Details
enQuireBio™
QP10849-20MG
20 mg
137.00€
20mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15915358
View Documents Promotion Details
enQuireBio™
QP10849-100MG
100 mg
212.00€
100mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925358
View Documents Promotion Details
enQuireBio™
QP10849-1GR
1 g
1450.00€
1g
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

PROK Tritirachium album Protein
< 1.0 EU per ug protein as determined by the LAL method.
34 Units/mg protein. One unit is defined as the amount of enzyme that will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 mol tyrosine per min at 37C, pH 7.5 (color by Folin-Ciocalteu reagent).
Other
Untagged
The Proteinase-K was lyophilized without any additives.
Research Use Only
PROK
Recombinant Protein
Yeast
MRLSVLLSLL PLALGAPAVE QRSEAAPLIE ARGEMVANKY IVKFKEGSAL SALDAAMEKI SGKPDHVYKN VFSGFAATLD ENMVRVLRAH PDVEYIEQDA VVTINAAQTN APWGLARISS TSPGTSTYYY DESAGQGSCV YVIDTGIEAS HPEFEGRAQM VKTYYYSSRD GNGHGTHCAG TVGSRTYGVA KKTQLFGVKV LDDNGSGQYS TIIAGMDFVA SDKNNRNCPK GVVASLSLGG GYSSSVNSAA ARLQSSGVMV AVAAGNNNAD ARNYSPASEP SVCTVGASDR YDRRSSFSNY GSVLDIFGPG TSILSTWIGG STRSISGTSM ATPHVAGLAA YLMTLGKTTA ASACRYIADT ANKGDLSNIP FGTVNLLAYN NYQA
Greater than 99% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications