missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other protein 1

A cDNA sequence encoding the Protein A was constructed and used to recombinantly synthesize the protein.

137.00€ - 2120.00€

Specifications

Name protein 1
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol spa
Biological Activity 7 mg human IgG/mg protein-A.
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15955368
View Documents Promotion Details
enQuireBio™
QP10853-EC-10MG
10 mg
137.00€
10mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15945368
View Documents Promotion Details
enQuireBio™
QP10853-EC-100MG
100 mg
545.00€
100mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15935368
View Documents Promotion Details
enQuireBio™
QP10853-1GRAM
1 g
2120.00€
1g
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

protein 1
< 1.0 EU per ug protein as determined by the LAL method.
7 mg human IgG/mg protein-A.
Other
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution containing no additives.
Research Use Only
spa
Recombinant Protein
E. coli
MKKKNIYSIR KLGVGIASVT LGTLLISGGV TPAANAAQHD EAQQNAFYQV LNMPNLNADQ RNGFIQSLKD DPSQSANVLG EAQKLNDSQA PKADAQQNKF NKDQQSAFYE ILNMPNLNEE QRNGFIQSLK DDPSQSTNVL GEAKKLNESQ APKADNNFNK EQQNAFYEIL NMPNLNEEQR NGFIQSLKDD PSQSANLLAE AKKLNESQAP KADNKFNKEQ QNAFYEILHL PNLNEEQRNG FIQSLKDDPS QSANLLAEAK KLNDAQAPKA DNKFNKEQQN AFYEILHLPN LTEEQRNGFI QSLKDDPSVS KEILAEAKKL NDAQAPKEED NNKPGKEDGN KPGKEDGNKP GKEDNKKPGK EDGNKPGKED NKKPGKEDGN KPGKEDGNKP GKEDGNKPGK EDGNKPGKED GNGVHVVKPG DTVNDIAKAN GTTADKIAAD NKLADKNMIK PGQELVVDKK QPANHADANK AQALPETGEE NPFIGTTVFG GLSLALGAAL LAGRRREL
Greater than 98.0% as determined by SDS-PAGE & RP-HPLC.
Videos
SDS
Documents

Documents

Product Certifications