missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other protein 1

Product Code. 15935368
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
This item is not returnable. View return policy

Product Code. 15935368

Brand: enQuireBio™ QP108531gram

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Protein A was constructed and used to recombinantly synthesize the protein.

Specifications

Name protein 1
Quantity 1 g
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol spa
Biological Activity 7 mg human IgG/mg protein-A.
Product Type Recombinant Protein
Cross Reactivity Other
Species E. coli
Protein Tag Untagged
Sequence MKKKNIYSIR KLGVGIASVT LGTLLISGGV TPAANAAQHD EAQQNAFYQV LNMPNLNADQ RNGFIQSLKD DPSQSANVLG EAQKLNDSQA PKADAQQNKF NKDQQSAFYE ILNMPNLNEE QRNGFIQSLK DDPSQSTNVL GEAKKLNESQ APKADNNFNK EQQNAFYEIL NMPNLNEEQR NGFIQSLKDD PSQSANLLAE AKKLNESQAP KADNKFNKEQ QNAFYEILHL PNLNEEQRNG FIQSLKDDPS QSANLLAEAK KLNDAQAPKA DNKFNKEQQN AFYEILHLPN LTEEQRNGFI QSLKDDPSVS KEILAEAKKL NDAQAPKEED NNKPGKEDGN KPGKEDGNKP GKEDNKKPGK EDGNKPGKED NKKPGKEDGN KPGKEDGNKP GKEDGNKPGK EDGNKPGKED GNGVHVVKPG DTVNDIAKAN GTTADKIAAD NKLADKNMIK PGQELVVDKK QPANHADANK AQALPETGEE NPFIGTTVFG GLSLALGAAL LAGRRREL
Buffer The protein was lyophilized from a concentrated (1 mg/ml) solution containing no additives.
Purity or Quality Grade Greater than 98.0% as determined by SDS-PAGE & RP-HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.