missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other RNASEH1 Protein
A cDNA sequence encoding the RNASEH1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | RNASEH1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933219
|
enQuireBio™
QP13325-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923219
|
enQuireBio™
QP13325-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913219
|
enQuireBio™
QP13325-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RNASEH1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
RNASEH1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 2mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLKQVEI FTDGSCLGNP GPGGYGAILR YRGREKTFSA GYTRTTNNRM ELMAAIVALE ALKEHCEVIL STDSQYVRQG ITQWIHNWKK RGWKTADKKP VKNVDLWQRL DAALGQHQIK WEWVKGHAGH PENERCDELA RAAAMNPTLE DTGYQVEV | |
Greater than 95% as determined by SDS-PAGE. |