missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant other SecB Protein

A cDNA sequence encoding the SecB was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Name SecB Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Other
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15933599
View Documents Promotion Details
enQuireBio™
QP13448-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15923599
View Documents Promotion Details
enQuireBio™
QP13448-25UG
25 μg
212.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15913599
View Documents Promotion Details
enQuireBio™
QP13448-1MG
1 mg
4090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

SecB Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Other
Untagged
The SecB protein solution (1 mg/ml) contains 20mM Tris-HCl pH 8 and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MSEQNNTEMT FQIQRIYTKD ISFEAPNAPH VFQKDWQPEV KLDLDTASSQ LADDVYEVVL RVTVTASLGE ETAFLCEVQQ GGIFSIAGIE GTQMAHCLGA YCPNILFPYA RECITSMVSR GTFPQLNLAP VNFDALFMNY LQQQAGEGTE EHQDA
Greater than 95.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications