Learn More
enQuireBio™ Recombinant other Sincalide Protein
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
Specifications
Specifications
| Name | Sincalide Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Other |
| Species | E. coli |
| Protein Tag | Untagged |
| Buffer | The Sincalide was lyophilized from a concentrated (1 mg/ml) solution with no additives. |
| Purity or Quality Grade | Greater than 99.0% as determined by RP-HPLC. |