missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Sincalide Protein
Recombinant Protein
137.00€ - 755.00€
Specifications
Name | Sincalide Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15913789
|
enQuireBio™
QP13506-1MG |
1 mg |
137.00€
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933789
|
enQuireBio™
QP13506-5MG |
5 mg |
212.00€
5mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923789
|
enQuireBio™
QP13506-25MG |
25 mg |
755.00€
25mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Sincalide Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 99.0% as determined by RP-HPLC. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The Sincalide was lyophilized from a concentrated (1 mg/ml) solution with no additives. |