missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other T. pallidum p17 (Partial) Protein
A cDNA sequence encoding the T. pallidum p17 (Partial) was constructed and used to recombinantly synthesize the protein.
238.00€ - 1515.00€
Specifications
Name | T. pallidum p17 (Partial) Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15914339
|
enQuireBio™
QP13672-100UG |
100 μg |
238.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934339
|
enQuireBio™
QP13672-500UG |
500 μg |
910.00€
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15924339
|
enQuireBio™
QP13672-1200UG |
1200 μg |
1515.00€
1200µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
T. pallidum p17 (Partial) Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
His | |
Treponema Pallidum protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
70mM Tris-HCl pH 8.0, 50mM NaCl, 50% Glycerol, 1.5M Urea. |